Anti-CAMK2A Goat Polyclonal Antibody
EVBIEB09376
- Antibody type:Primary
- Antigen name:calcium/calmodulin-dependent Protein kinase II alpha
- Antigen symbol:CAMK2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CAMK2A (calcium/calmodulin-dependent protein kinase II alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat