Anti-AGAP2 Goat Polyclonal Antibody
Catalog # EVBIEB09314
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
- Antigen symbol:AGAP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: AGAP2 (ArfGAP with GTPase domain, ankyrin repeat and PH domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human