Anti-KCNJ1 / ROMK Goat Polyclonal Antibody
EVBIEB09130
- Antibody type:Primary
- Antigen symbol:KCNJ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Antigen synonyms:ROMK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: KCNJ1 / ROMK
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat