Anti-GAD1 (isoform GAD67) Goat polyclonal antibody unconjugated
EVBIEB09109
- Antibody type:Primary
- Antigen name:GAD1 (isoform GAD67)
- Antigen symbol:GAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: GAD1 (isoform GAD67)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: