Anti-PSCA Goat Polyclonal Antibody
EVBIEB08994
- Antibody type:Primary
- Antigen name:Prostate Stem Cell Antigen
- Antigen symbol:PSCA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Amino acid number:27 to 40
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: PSCA (prostate stem cell antigen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human