Anti-CHRNA7 Goat polyclonal antibody unconjugated
EVBIEB08853
- Antibody type:Primary
- Antigen name:cholinergic receptor, nicotinic, alpha 7 (neuronal)
- Antigen symbol:CHRNA7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: CHRNA7 (cholinergic receptor, nicotinic, alpha 7 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: