Order Entry
Austria
ContactUsLinkComponent
 
Anti-FFAR4 Goat Polyclonal Antibody
  EVBIEB08837
Anti-FFAR4 Goat Polyclonal Antibody
  EVBIEB08837

 

  • Antibody type:
    Primary
  • Antigen name:
    Free fatty acid receptor 4
  • Antigen symbol:
    FFAR4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: FFAR4 (free fatty acid receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat