Anti-SLC10A2 / ASBT Goat Polyclonal Antibody
Catalog # EVBIEB08818
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen symbol:Solute carrier family 10, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:C-terminal
- Cross adsorption:none
- Antigen synonyms:ASBT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SLC10A2 / ASBT
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human