Anti-PSD3 Goat Polyclonal Antibody
EVBIEB08278
- Antibody type:Primary
- Antigen name:pleckstrin and Sec7 domain containing 3
- Antigen symbol:PSD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Amino acid number:327 to 339
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: PSD3 (pleckstrin and Sec7 domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human