Anti-APOBEC3D Goat polyclonal antibody unconjugated
EVBIEB08270
- Antibody type:Primary
- Antigen name:Apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3D
- Antigen symbol:APOBEC3D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: APOBEC3D (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human