Order Entry
Austria
ContactUsLinkComponent
Anti-PGBD1 Goat Polyclonal Antibody
Anti-PGBD1 Goat Polyclonal Antibody
Catalog # EVBIEB08242
Supplier:  Everest Biotech
Anti-PGBD1 Goat Polyclonal Antibody
Catalog # EVBIEB08242
Supplier:  Everest Biotech

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PiggyBac Transposable Element Derived 1
  • Antigen symbol:
    PGBD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: PGBD1 (piggyBac transposable element derived 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human