Anti-MAN2A1 Goat Polyclonal Antibody
EVBIEB08228
- Antibody type:Primary
- Antigen name:Mannosidase, alpha, class 2A, member 1
- Antigen symbol:MAN2A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: MAN2A1 (mannosidase, alpha, class 2A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human