Anti-NLRP3 Goat Polyclonal Antibody
EVBIEB07335
- Antibody type:Primary
- Antigen name:NLR family, pyrin domain containing 3
- Antigen symbol:NLRP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: NLRP3 (NLR family, pyrin domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: