Anti-PIAS2 Goat Polyclonal Antibody
EVBIEB07176
- Antibody type:Primary
- Antigen name:Protein inhibitor of activated STAT 2
- Antigen symbol:PIAS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: PIAS2 (protein inhibitor of activated STAT, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat