Anti-VPS25 Goat Polyclonal Antibody
EVBIEB07071
- Antibody type:Primary
- Antigen name:Vacuolar Protein Sorting 25
- Antigen symbol:VPS25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: VPS25 (vacuolar protein sorting 25 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat