Anti-EPM2A Goat Polyclonal Antibody
Catalog # EVBIEB06610
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Epilepsy progressive myoclonus type 2A Lafora disease (laforin)
- Antigen symbol:EPM2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: EPM2A (epilepsy, progressive myoclonus type 2A, Lafora disease (laforin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: