Anti-IFT74 Goat Polyclonal Antibody
EVBIEB06339
- Antibody type:Primary
- Antigen name:intraflagellar transport 74 homolog (Chlamydomonas)
- Antigen symbol:IFT74
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: IFT74 (intraflagellar transport 74 homolog (Chlamydomonas))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Rat