Anti-LIN7B / MALS-2 Goat Polyclonal Antibody
EVBIEB06205
- Antibody type:Primary
- Antigen symbol:LIN7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Antigen synonyms:MALS-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: LIN7B / MALS-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: