Anti-OSBPL1A Goat Polyclonal Antibody
Artikelnr. EVBIEB06151
Lieferant: Everest Biotech
Spezifikationen
- Antibody type:Primary
- Antigen name:oxysterol binding Protein-like 1A
- Antigen symbol:OSBPL1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Spezifikationen
Über diesen Artikel
Type: Primary
Antigen: OSBPL1A (oxysterol binding protein-like 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: