Anti-MRPL3 Goat Polyclonal Antibody
EVBIEB05689
- Antibody type:Primary
- Antigen name:mitochondrial ribosomal Protein L3
- Antigen symbol:MRPL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Type: Primary
Antigen: MRPL3 (mitochondrial ribosomal protein L3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: