Anti-Gli2 Rabbit Polyclonal Antibody
Catalog # BSBTPA1941
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:GLI family zinc finger 2
- Antigen symbol:GLI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:THP2|HPE9|CJS|PHS2|THP1
- Amino acid number:1444 to 1463
- Sequence:MLYYYGQIHMYEQDGGLENL
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Gli2 (1444-1463aa MLYYYGQIHMYEQDGGLENL).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Zinc finger protein GLI2(GLI2) detection. Tested with WB in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Gli2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human