Anti-PKB alpha Mouse Monoclonal Antibody [clone: IML-26]
Catalog # BSBTMA1085
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:PKB alpha
- Antigen symbol:PKBA
- Clonality:Monoclonal
- Clone:IML-26
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Amino acid number:461 to 477
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Synthetic peptide corresponding to amino acids 461-477 of human PKB alpha/Akt1, conjugated to KLH.
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for PKB alpha, v-akt murine thymoma viral oncogene homolog 1 (AKT1) detection. Tested with WB in Human; mouse; rat; chicken. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: PKB alpha
Clonality: Monoclonal
Clone: IML-26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken