Anti-H2AFX Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27428
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:H2A histone family, member X
- Antigen symbol:H2AFX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:3014
- Antigen synonyms:H2A/X|H2A.X|H2AX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: H2AFX (H2A histone family, member X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human