Anti-Blimp-1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-27180
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:PR domain containing 1, with ZNF domain
- Antigen symbol:PRDM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:639
- Antigen synonyms:PRDI-BF1|BLIMP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Blimp-1 (Blimp-1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human