Anti-TAAR1 Rabbit Polyclonal Antibody
NOVUNBP2-24720
- Antibody type:Primary
- Antigen name:Trace amine associated receptor 1
- Antigen symbol:TAAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Gene ID:134864
- Antigen synonyms:TRAR1|TA1|TAR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: TAAR1 (trace amine associated receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: