Anti-FURIN Rabbit Polyclonal Antibody
NOVUNBP2-24687
- Antibody type:Primary
- Antigen name:Furin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Gene ID:5045
- Antigen synonyms:SPC1|FUR|PACE|PCSK3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: FURIN (furin (paired basic amino acid cleaving enzyme))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: