Anti-ZEB1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ZEB1 (zinc finger E-box binding homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Specifications
- Catalog No:
- NOVUNBP2-13159
- NOVUNBP2-13159SS
- Antigen symbol:
- ZEB1
- ZEB1
- Antigen name:
- Zinc Finger E-box Binding Homeobox 1
- Zinc Finger E-box Binding Homeobox 1
- Antigen synonyms:
- ZFHEP|TCF8|FECD6|PPCD3|ZFHX1A|NIL2A|TCF-8|NIL-2-A zinc finger protein|DELTAEF1|BZP|AREB6|Zinc finger E-box-binding homeobox 1
- ZFHEP|TCF8|FECD6|PPCD3|ZFHX1A|NIL2A|TCF-8|NIL-2-A zinc finger protein|DELTAEF1|BZP|AREB6|Zinc finger E-box-binding homeobox 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6935
- 6935
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together