Anti-GABARAP Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The GABARAP Antibody from Novus Biologicals is a rabbit polyclonal antibody to GABARAP. This antibody reacts with mouse. The GABARAP Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC / IF (Negative).
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: GABARAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- NOVUNBP1-97416
- NOVUNBP1-97416SS
- Antigen symbol:
- GABARAP
- GABARAP
- Antigen name:
- Gamma-aminobutyric acid receptor-associated protein
- Gamma-aminobutyric acid receptor-associated protein
- Antigen synonyms:
- GABARAP-a|MM46|GABA(A) receptor-associated protein|FLC3B|ATG8A|Gamma-aminobutyric acid receptor-associated protein
- GABARAP-a|MM46|GABA(A) receptor-associated protein|FLC3B|ATG8A|Gamma-aminobutyric acid receptor-associated protein
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 11337
- 11337
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together