Anti-NLRC4 Rabbit Polyclonal Antibody
The CARD12 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CARD12. This antibody reacts with human, mouse. The CARD12 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: CARD12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- NOVUNBP1-78980
- NOVUNBP1-78980SS
- Antigen symbol:
- CARD12
- CARD12
- Antigen name:
- Caspase Recruitment Domain Family, Member 12
- Caspase Recruitment Domain Family, Member 12
- Antigen synonyms:
- CLANA|AIFEC|CLAN|FCAS4|CARD12|CLAN1|CLR2.1|CLANC|IPAF|CLAND|CLANB
- CLANA|AIFEC|CLAN|FCAS4|CARD12|CLAN1|CLR2.1|CLANC|IPAF|CLAND|CLANB
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 58484
- 58484
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together