Order Entry
Austria
ContactUsLinkComponent
Anti-PIEZO2 Rabbit Polyclonal Antibody (DyLight® 550)
Anti-PIEZO2 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-78538R
Supplier:  Novus Biologicals
Anti-PIEZO2 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNBP1-78538R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Piezo-type Mechanosensitive Ion Channel Component 2
  • Antigen symbol:
    PIEZO2
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    63895
  • Antigen synonyms:
    HsT771|C18orf58|FLJ23144|DA5|HsT748|C18orf30|FAM38B|FLJ23403|FAM38B2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: PIEZO2 (piezo-type mechanosensitive ion channel component 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse