Order Entry
Austria
ContactUsLinkComponent
Anti-ABCB5 Rabbit Polyclonal Antibody
Anti-ABCB5 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-ABCB5 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-77687SS
  • NOVUNBP1-77687
  • Antigen symbol:
  • ABCB5
  • ABCB5
  • Antigen name:
  • ATP Binding Cassette, Subfamily B, Member 5
  • ATP Binding Cassette, Subfamily B, Member 5
  • Antigen synonyms:
  • P-glycoprotein ABCB5|ATP-binding cassette sub-family B member 5|ABCB5 P-gp|ABCB5
  • P-glycoprotein ABCB5|ATP-binding cassette sub-family B member 5|ABCB5 P-gp|ABCB5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 340273
  • 340273
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together