Anti-TRPV1 Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-71774V
- Antibody type:Primary
- Antigen name:transient receptor potential cation channel, subfamily V, member 1
- Antigen symbol:TRPV1
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Gene ID:7442
- Antigen synonyms:VR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: TRPV1 (transient receptor potential cation channel, subfamily V, member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse