Anti-GABARAP Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNBP1-71771V
- Antibody type:Primary
- Antigen name:GABA(A) Receptor-Associated Protein
- Antigen symbol:GABARAP
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:11337
- Antigen synonyms:GABARAP-a|MM46|ATG8A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: GABARAP (GABA(A) receptor-associated protein)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse