Anti-GABARAP Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-71771G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid receptor-associated protein
- Antigen symbol:GABARAP
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Gene ID:11337
- Antigen synonyms:GABARAP-a|MM46|GABA(A) receptor-associated protein|FLC3B|ATG8A|Gamma-aminobutyric acid receptor-associated protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: GABARAP (GABA(A) receptor-associated protein)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse