Order Entry
Austria
ContactUsLinkComponent
Anti-SDHB Rabbit Polyclonal Antibody
Anti-SDHB Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-SDHB Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-54154
  • NOVUNBP1-54154SS
  • Antigen symbol:
  • SDHB
  • SDHB
  • Antigen name:
  • Succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
  • Succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
  • Antigen synonyms:
  • SDHIP|CWS2|IP|PGL4|SDH|SDH1|SDH2
  • SDHIP|CWS2|IP|PGL4|SDH|SDH1|SDH2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 6390
  • 6390
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together