Anti-RICTOR Mouse Monoclonal Antibody [clone: 7B3]
Supplier: Novus Biologicals
Certificates
About this item
The Rictor Antibody (7B3) from Novus Biologicals is a mouse monoclonal antibody to Rictor. This antibody reacts with human, mouse, primate. The Rictor Antibody (7B3) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000
Type: Primary
Antigen: Rictor
Clonality: Monoclonal
Clone: 7B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Primate
Specifications
- Catalog No:
- NOVUNBP1-51645
- NOVUNBP1-51645SS
- Antigen symbol:
- RICTOR
- RICTOR
- Antigen name:
- Rapamycin-insensitive companion of mTOR
- RPTOR independent companion of MTOR, complex 2
- Antigen synonyms:
- AVO3 homolog|AVO3|PIA|RICTOR|Rapamycin-insensitive companion of mTOR|KIAA1999|hAVO3
- AVO3|PIA|hAVO3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 7B3
- 7B3
- Reactivity:
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 253260
- 253260
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together