Anti-MAP1B Rabbit Polyclonal Antibody (DyLight® 650) [clone: SuperBUGS]
Catalog # NOVUNBP1-42827C
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Microtubule-associated protein 1B
- Antigen symbol:MAP1B
- Clonality:Polyclonal
- Clone:SuperBUGS
- Conjugation:DyLight® 650
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:4131
- Antigen synonyms:MAP-1B|MAP5|PPP1R102|Microtubule-associated protein 1B|MAP1B|FUTSCH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: MAP1B (microtubule-associated protein 1B)
Clonality: polyclonal
Clone: SuperBUGS
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: