Anti-AGT Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNBP1-30027R
- Antibody type:Primary
- Antigen name:Angiotensin
- Antigen symbol:AGT
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:183
- Antigen synonyms:Ang II|Angiotensin-3|Angiotensin I|Des-Asp[1]-angiotensin II|Angiotensin-2|SERPINA14|Ang I|Angiotensinogen|Angiotensin-1|Angiotensin II|Ang III|Serpin A8|Angiotensin III|AGT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µL
- Pk:0,2 mL
Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat