Anti-BMP2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-19751F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:bone morphogenetic protein 2
- Antigen symbol:BMP2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:650
- Antigen synonyms:BMP2A|BDA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: BMP2 (bone morphogenetic protein 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat