Anti-TCF7L2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
NOVUNBP1-19083F
- Antibody type:Primary
- Antigen name:Transcription factor 7-like 2
- Antigen symbol:TCF7L2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:6934
- Antigen synonyms:TCF4|TCF-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: TCF7L2 (transcription factor 7-like 2 (T-cell specific, HMG-box))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human