Anti-TCF7L2 Rabbit Polyclonal Antibody (Biotin)
NOVUNBP1-19083B
- Antibody type:Primary
- Antigen name:Transcription factor 7-like 2
- Antigen symbol:TCF7L2
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:6934
- Antigen synonyms:TCF7L2|TCF4|Transcription factor 7-like 2|T-cell factor 4|TCF-4|T-cell-specific transcription factor 4|HMG box transcription factor 4|hTCF-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: TCF7L2 (transcription factor 7-like 2 (T-cell specific, HMG-box))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human