Order Entry
Austria
ContactUsLinkComponent
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2F3]
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2F3]
Supplier:  Novus Biologicals
Certificates
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-2F3]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-18964
  • NOVUNBP1-18964SS
  • Antigen name:
  • Oval Cell Marker
  • Oval Cell Marker
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • OC2-2F3
  • OC2-2F3
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together