Anti-CCL2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNBP1-07034F
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 2
- Antigen symbol:CCL2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:6347
- Antigen synonyms:HC11|MCAF|SMC-CF|GDCF-2|HSMCR30|SCYA2|MCP1|MCP-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat