Order Entry
Austria
ContactUsLinkComponent
 
Anti-LDL Receptor Rabbit Polyclonal Antibody
 
undefined
Anti-LDL Receptor Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNBP1-06709SS
  • NOVUNBP1-06709
  • Antigen symbol:
  • LDLR
  • LDLR
  • Antigen name:
  • Low-Density Lipoprotein Receptor
  • Low-Density Lipoprotein Receptor
  • Antigen synonyms:
  • FHC|FH|LDLCQ2|LDL receptor|LDL-R|Low-density lipoprotein receptor
  • FHC|FH|LDLCQ2|LDL receptor|LDL-R|Low-density lipoprotein receptor
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 3949
  • 3949
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together