Order Entry
Austria
ContactUsLinkComponent
Anti-DIO3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-DIO3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  NOVUNBP1-05767F
Anti-DIO3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
  NOVUNBP1-05767F

 

  • Antibody type:
    Primary
  • Antigen name:
    deiodinase, iodothyronine, type III
  • Antigen symbol:
    DIO3
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    1735
  • Antigen synonyms:
    TXDI3|DIOIII|D3|5DIII
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: DIO3 (deiodinase, iodothyronine, type III)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: