Order Entry
Austria
ContactUsLinkComponent
Anti-Blimp-1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 3H2-E8]
Anti-Blimp-1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 3H2-E8]
Catalog # NOVUNB600-235C
Supplier:  Novus Biologicals
undefined
Anti-Blimp-1 Mouse Monoclonal Antibody (DyLight® 650) [clone: 3H2-E8]
Catalog # NOVUNB600-235C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PR Domain Zinc Finger Protein 1
  • Antigen symbol:
    BLIMP1
  • Clonality:
    Monoclonal
  • Clone:
    3H2-E8
  • Conjugation:
    DyLight® 650
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    639
  • Antigen synonyms:
    PRDI-BF1|B-lymphocyte-induced maturation protein 1|PR domain zinc finger protein 1|RP1-134E15.1|BLIMP1|BLIMP-1|PRDI-binding factor-1|beta-interferon gene positive-regulatory domain I binding factor
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

The BLIMP1 / PRDM1 Antibody (3H2-E8) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to BLIMP1 / PRDM1. This antibody reacts with human, mouse, porcine. The BLIMP1 / PRDM1 Antibody (3H2-E8) [DyLight 650] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: BLIMP1/PRDM1
Clonality: Monoclonal
Clone: 3H2-E8
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Porcine