Order Entry
Austria
ContactUsLinkComponent
 
Anti-CASP7 Mouse monoclonal antibody unconjugated [clone: Mch3 1-1-11]
 
undefined
Anti-CASP7 Mouse monoclonal antibody unconjugated [clone: Mch3 1-1-11]

 

  • Catalog No:
  • NOVUNB500-206SS
  • NOVUNB500-206
  • Antigen symbol:
  • CASP7
  • CASP7
  • Antigen name:
  • Caspase 7
  • Caspase 7
  • Antigen synonyms:
  • Caspase 7 Apoptosis-related Cysteine Peptidase|CASP-7|MCH3|CASP7|ICE-LAP3|CMH-1
  • Caspase 7 Apoptosis-related Cysteine Peptidase|CASP-7|MCH3|CASP7|ICE-LAP3|CMH-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Mch3 1-1-11
  • Mch3 1-1-11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 840
  • 840
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

Frequently Bought Together