Anti-ABCE1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: ABCE1 (ATP-binding cassette, sub-family E (OABP), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB400-116
- NOVUNB400-116SS
- Antigen symbol:
- ABCE1
- ABCE1
- Antigen name:
- ATP Binding Cassette, Subfamily E, Member 1
- ATP Binding Cassette, Subfamily E, Member 1
- Antigen synonyms:
- RNase 4 inhibitor|Ribonuclease 4 Inhibitor|ABC 38|ABCE 1|OABP|ATP Binding Cassette Sub Family E Member 1|RLI|RNS4I|ABC38|RNase L Inhibitor
- RNase 4 inhibitor|Ribonuclease 4 Inhibitor|ABC 38|ABCE 1|OABP|ATP Binding Cassette Sub Family E Member 1|RLI|RNS4I|ABC38|RNase L Inhibitor
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 6059
- 6059
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together