Order Entry
Austria
ContactUsLinkComponent
Anti-SCARB1 Rabbit Polyclonal Antibody
Anti-SCARB1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
Anti-SCARB1 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB400-113
  • NOVUNB400-113SS
  • Antigen symbol:
  • SCARB1
  • SCARB1
  • Antigen name:
  • Scavenger receptor class B, member 1
  • Scavenger receptor class B, member 1
  • Antigen synonyms:
  • SR-BI|HDLQTL6|SRB1|CLA-1|CD36L1|CLA1
  • SR-BI|HDLQTL6|SRB1|CLA-1|CD36L1|CLA1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 949
  • 949
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together