Order Entry
Austria
ContactUsLinkComponent
Anti-GRM6 Rabbit Polyclonal Antibody
Anti-GRM6 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-GRM6 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB300-189SS
  • NOVUNB300-189
  • Antigen symbol:
  • GRM6
  • GRM6
  • Antigen name:
  • Glutamate Receptor, Metabotropic 6
  • Glutamate Receptor, Metabotropic 6
  • Antigen synonyms:
  • GPRC1F|CSNB1B|mGlu6|MGLUR6
  • GPRC1F|CSNB1B|mGlu6|MGLUR6
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 2916
  • 2916
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together